Polypropylene String-Wound Gradient-Density Cartridges
String-wind pattern has a lower presure drop for higher flow
Ideal for removing a broad range of deformable and nondeformable particles such as sand, silt, sludge, rust, and scale. Polypropylene string is resistant to chemical and bacterial attack. Maximum temperature is 150°F (66°C).
Note: For optimal performance, change cartridge at 25 psid.
$11.00 - $16.10USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Length (in) Flow Rate Micron Rating
Availability Pricing
COMPARE
10 7 GPM 5
$14.20USD / Each
COMPARE
10 10 GPM 30
$11.00USD / Each
COMPARE
10 10 GPM 50
$16.10USD / Each