PolyScience DuraChill® Portable Recirculating Chillers
Maximize airflow and reduce maintenance with self-cleaning air filter
  • Continuous fluid level monitoring protects pump and prevents freezing
  • Easy access to front fill reservoir port saves time and avoids spills
  • Noise reduction system creates quieter work environment
  • Fluid path stays clean without chemical additives
  • Large color touch screen displays key information at a glance
  • USB port allows for data logging
Available with either a turbine or positive displacement pump, these portable PolyScience DuraChill recirculating chillers feature a modulated refrigeration system designed to use less refrigerant, enhance temperature stability, extend compressor life, and allow you to migrate to natural refrigerants with little or no global warming effects.
Self-changing air filter system automatically changes the filter media at user-definable intervals or once a month for a two-year period helping to reduce downtime and maintenance. An easy-to-read large full-color touchscreen display offers a multilanguage interface and shows set temperature, actual fluid temperature, fluid pressure, fluid level, air filter status, and timer.
State-of-the-art capacitance liquid level sensor provides continuous monitoring to protect the pump from premature failure. A special noise reduction system makes the chillers extremely quiet while still maintaining optimal temperature control. A UV light biological growth inhibitor eliminates the need for chemical growth inhibitors and prevents algicides from being released into the waterways. Self-diagnostic system allows you to troubleshoot performance issues against data stored on the chiller the day it was manufactured.
$4,645.00 - $6,895.00USD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cooling Capacity At 0° C (watts) Max Flow Rate (L/min) Power (VAC)
Availability Pricing
PolyScience Durachill CA02 Recirculating Chiller with Positive Displacement Pump, 1/4HP, 240V, 50Hz EW-12930-01 Mfr # CA02A2P1-41AA1N
COMPARE
300 7.6 240
$4,645.00USD / Each
PolyScience Durachill CA02 Recirculating Chiller with Positive Displacement Pump, 1/4HP, 120V, 60Hz EW-12930-02 Mfr # CA02A1P1-41AA1N
COMPARE
400 9.8 120
$4,645.00USD / Each
PolyScience Durachill CA02 Recirculating Chiller with Turbine Pump, 1/4HP, 240V, 50Hz EW-12930-03 Mfr # CA02A2T1-41AA1N
COMPARE
300 8.3 240
$4,990.00USD / Each
PolyScience Durachill CA02 Recirculating Chiller with Turbine Pump, 1/4HP, 120V, 60Hz EW-12930-04 Mfr # CA02A1T1-41AA1N
COMPARE
400 13.2 120
$4,990.00USD / Each
PolyScience Durachill CA03 Recirculating Chiller with Turbine Pump, 1/3HP, 240V, 50Hz EW-12930-06 Mfr # CA03A2T1-41AA1N
COMPARE
490 8.3 240
$5,335.00USD / Each
PolyScience Durachill CA03 Recirculating Chiller with Turbine Pump, 1/3HP, 120V, 60Hz EW-12930-07 Mfr # CA03A1T1 - 41AA1N
COMPARE
530 13.2 120
$5,335.00USD / Each
PolyScience Durachill CA03 Recirculating Chiller with Positive Displacement Pump, 1/3HP, 240V, 50Hz EW-12930-08 Mfr # CA03A2P1-41AA1N
COMPARE
490 7.6 240
$4,960.00USD / Each
PolyScience Durachill CA03 Recirculating Chiller with Positive Displacement Pump, 1/3HP, 120V, 60Hz EW-12930-09 Mfr # CA03A1P1-41AA1N
COMPARE
530 9.8 120
$4,960.00USD / Each
PolyScience Durachill CA05 Recirculating Chiller with Turbine Pump, 1/2HP, 120V, 60Hz EW-12930-11 Mfr # CA05A1T1-41AA1N
COMPARE
770 13.2 120
$5,665.00USD / Each
PolyScience Durachill CA05 Recirculating Chiller with Turbine Pump, 1/2HP, 240V, 50Hz EW-12930-13 Mfr # CA05A2T1-41AA1N
COMPARE
770 8.3 240
$5,665.00USD / Each
PolyScience Durachill CA05 Recirculating Chiller with Positive Displacement Pump, 1/2HP, 120V, 60Hz EW-12930-14 Mfr # CA05A1P1-41AA1N
COMPARE
770 9.8 120
$5,305.00USD / Each
PolyScience Durachill CA05 Recirculating Chiller with Positive Displacement Pump, 1/2HP, 240V, 50Hz EW-12930-16 Mfr # CA05A2P1-41AA1N
COMPARE
770 7.6 240
$5,305.00USD / Each
PolyScience Durachill CA10 Recirculating Chiller with Positive Displacement Pump, 1HP, 208-240V, 60Hz EW-12930-17 Mfr # CA10A3P2-41AA1N
COMPARE
1000 13.2 208 / 240
$6,545.00USD / Each
PolyScience Durachill CA10 Recirculating Chiller with Positive Displacement Pump, 1HP, 240V, 50Hz EW-12930-18 Mfr # CA10A2P2-41AA1N
COMPARE
1000 11 240
$6,545.00USD / Each
PolyScience Durachill CA10 Recirculating Chiller with Turbine Pump, 1HP, 208-240V, 60Hz EW-12930-19 Mfr # CA10A3T1-41AA1N
COMPARE
1000 13.2 208 / 240
$6,895.00USD / Each
PolyScience Durachill CA10 Recirculating Chiller with Turbine Pump, 1HP, 240V, 50Hz EW-12930-21 Mfr # CA10A2T1-41AA1N
COMPARE
1000 11 240
$6,895.00USD / Each
Showing 1- 16 of 16