This product is now discontinued, View Related Products

PolyScience Heated Circulating Baths

High-performance units with the heating features you need
  • All units feature over-temperature and low-level cutoffs to protect you and your samples
  • Space-saving design minimizes bench space utilization while providing excellent performance
These quality baths are ideal for calibration, melting curves, enzyme assays, temperature stability studies, or other applications requiring precise heating.
Call For Price

20 variations of this product are available.

See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Reservoir Capacity (L)
Availability Pricing
PolyScience 80A0A11B 6-liter Analog Controller Heated Circulating Bath, 120VAC EW-12116-00 Mfr # 80A0A11B
COMPARE
150 Ambient 27
Call For Price
PolyScience 80A0A12E 6-liter Analog Controller Heated Circulating Bath, 240VAC EW-12116-05 Mfr # 80A0A12E
COMPARE
150 Ambient 27
Call For Price
PolyScience SD07H170 7L Standard Digital Heated Circulating Bath, 120VAC EW-12116-10 Mfr # SD07H170-A11B
COMPARE
170 Ambient +5 7
Call For Price
PolyScience 8006A12E 6L Standard Digital Heated Circulating Bath, 240VAC EW-12116-15 Mfr # 8006A12E
COMPARE
150 Ambient +5 10
Call For Price
PolyScience 8002A11B 6-liter Advanced Digital Controller Heated Circulating Bath, 120VAC EW-12116-20 Mfr # 8002A11B
COMPARE
150 Ambient 12
Call For Price
PolyScience 8002A12E 6-liter Advanced Digital Controller Heated Circulating Bath, 240VAC EW-12116-25 Mfr # 8002A12E
COMPARE
150 Ambient 12
Call For Price
PolyScience 8012A11B Programmable Heated 6L Circulating Bath, 120VAC/60Hz EW-12116-30 Mfr # 8012A11B
COMPARE
200 Ambient +5 10
Call For Price
PolyScience 8012A12E Programmable Heated 6L Circulating Bath, 240VAC/50Hz EW-12116-35 Mfr # 8012A12E
COMPARE
200 Ambient +5 10
Call For Price
PolyScience 8106A11B 13-liter Standard Digital Controller Heated Circulating Bath, 120VAC EW-12116-40 Mfr # 8106A11B
COMPARE
150 Ambient 12
Call For Price
PolyScience 8106A12E 13-liter Standard Digital Controller Heated Circulating Bath, 240VAC EW-12116-45 Mfr # 8106A12E
COMPARE
150 Ambient 12
Call For Price
PolyScience 8102A11B 13-liter Advanced Digital Controller Heated Circulating Bath, 120VAC EW-12116-50 Mfr # 8102A11B
COMPARE
150 Ambient 12
Call For Price
PolyScience 8102A12E 13-liter Advanced Digital Controller Heated Circulating Bath, 240VAC EW-12116-55 Mfr # 8102A12E
COMPARE
150 Ambient 12
Call For Price
PolyScience 8112A11B 13-liter Programmable Controller Heated Circulating Bath, 120VAC EW-12116-60 Mfr # 8112A11B
COMPARE
150 Ambient 4.5
Call For Price
PolyScience 8112A12E 13-liter Programmable Controller Heated Circulating Bath, 240VAC EW-12116-65 Mfr # 8112A12E
COMPARE
150 Ambient 4.5
Call For Price
PolyScience 8206A11B 28-liter Standard Digital Controller Heated Circulating Bath, 120VAC EW-12116-70 Mfr # 8206A11B
COMPARE
150 Ambient 8
Call For Price
PolyScience 8206A12E 28-liter Standard Digital Controller Heated Circulating Bath, 240VAC EW-12116-75 Mfr # 8206A12E
COMPARE
150 Ambient 28
Call For Price
PolyScience 8202A11B 28-liter Advanced Digital Controller Heated Circulating Bath, 120VAC EW-12116-80 Mfr # 8202A11B
COMPARE
150 Ambient 28
Call For Price
PolyScience 8202A12E 28-liter Advanced Digital Controller Heated Circulating Bath, 240VAC EW-12116-85 Mfr # 8202A12E
COMPARE
150 Ambient 28
Call For Price
PolyScience 8212A11B 28-liter Programmable Controller Heated Circulating Bath, 120VAC EW-12116-90 Mfr # 8212A11B
COMPARE
150 Ambient 28
Call For Price
PolyScience 8212A12E 28-liter Programmable Controller Heated Circulating Bath, 240VAC EW-12116-95 Mfr # 8212A12E
COMPARE
150 Ambient 28
Call For Price
Showing 1- 20 of 20

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat