PolyScience Histology Freeze Plate

Ultra-cold surface reduces tissue processing time by 40%
  • Monitor tissue specimen quality on the fast-freezing surface
  • Process multiple samples at once on the large 136 in2 freezing area
  • Achieve working temperature in only 10 to 15 minutes for quick start up
The histology freeze plate is a necessity for processing specimens for sectioning in a microtome with no cryostat, or to perform dissections of frozen tissue. It speeds up the curing process of tissue blocks and keeps frozen tissue from thawing, increasing productivity in the laboratory.
$2,885.00 - $3,125.00USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
PolyScience HPLAT-30-A11B Histology Freeze Plate, 120 VAC EW-48573-10 Mfr # HPLAT-30-A11B
COMPARE
115
$2,885.00USD / Each
PolyScience HPLAT-30-A12E Histology Freeze Plate, 240 VAC EW-48573-15 Mfr # HPLAT-30-A12E
COMPARE
240
$3,125.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat