PolyScience Stainless Steel Open Bath with MX Immersion Circulator Systems
  • Large user-friendly digital display
  • Integral heating coil and re-circulating pump
  • Stainless steel tank with drain valve, bridge and lid
Designed with a large, easy-to-read backlit display that easily shows both the process value and set value from a distance, as well as heat-on and circulating-on icons. Rear ports can be used for external process temperature control. The four pushbuttons allow quick control of the on/off/standby mode, set-point value, Celsius/Fahrenheit selection, and offset calibration value. The low/high limit settings control the min/max temperature, and the compact, stylish design has a streamlined profile to occupy minimum reservoir space. A protective housing prevents direct contact with the integral pump and heater coil, while offering easy access for cleaning and inspection. A convenient slide control allows easy fluid flow adjustment from the single-speed circulating pump.
$1,775.00 - $2,115.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Reservoir Capacity (L)
Availability Pricing
PolyScience MX06S135-A11B 6L, Open Bath System w/ Lid, Ambient +10 to 135 C; 120 VAC/60 Hz EW-12101-66 Mfr # MX06S135-A11B
COMPARE
115 6
$1,850.00USD / Each
PolyScience MX06S135-A12E 6L, Open Bath System w/ Lid, Ambient +10 to 135 C; 240 VAC/50 Hz EW-12101-67 Mfr # MX06S135-A12E
COMPARE
230 6
$1,850.00USD / Each
PolyScience MX10S135-A11B 10L, Open Bath System w/ Lid, Ambient +10 to 135 C; 120 VAC/60 Hz EW-12101-68 Mfr # MX10S135-A11B
COMPARE
115 10
$1,775.00USD / Each
PolyScience MX10S135-A12E 10L, Open Bath System w/ Lid, Ambient +10 to 135 C; 240 VAC/50 Hz EW-12101-69 Mfr # MX10S135-A12E
COMPARE
230 10
$1,775.00USD / Each
PolyScience MX20S135-A11B 20L, Open Bath System w/ Lid, Ambient +10 to 135 C; 120 VAC/60 Hz EW-12101-72 Mfr # MX20S135-A11B
COMPARE
115 20
$1,885.00USD / Each
PolyScience MX20S135-A12E 20L, Open Bath System w/ Lid, Ambient +10 to 135 C; 240 VAC/50 Hz EW-12101-73 Mfr # MX20S135-A12E
COMPARE
230 20
$1,930.00USD / Each
PolyScience MX28S135-A11B 28L, Open Bath System w/ Lid, Ambient +10 to 135 C; 120 VAC/60 Hz EW-12101-74 Mfr # MX28S135-A11B
COMPARE
115 28
$2,115.00USD / Each
PolyScience MX28S135-A12E 28L, Open Bath System w/ Lid, Ambient +10 to 135 C; 240 VAC/50 Hz EW-12101-75 Mfr # MX28S135-A12E
COMPARE
230 28
$2,115.00USD / Each
Showing 1- 8 of 8