Pomona Circular Connector Adapter Set
Available for sale only in the United States and Canada
  • Full set of pin and socket adapters
  • 18 piece set includes one black and red pin and socket of each size
  • Contact sizes: 0.03in, 0.04in, 0.06in, 0.10in
  • Includes Silicone insulated 4ft leads to fit most DMMs from Fluke, Amprobe, Greenlee and others
ITEM(S) ON SALE
$101.69 - $186.29USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Pomona Electronics 6481 Test Connector Adapter Set, 18 Pieces EW-20009-45 Mfr # 6481
COMPARE
$101.69USD / Each
$112.99USD / Each
Pomona Electronics 6730 KELVIN LEAD SETWIDE JAW EW-20009-58 Mfr # 6730
COMPARE
$186.29USD / Each
$206.99USD / Each