Pomona Mini Test Probe
Available for sale only in the United States and Canada.
SMD test probe with sharp, spring mounted tip and 2mm jack. Provides ultra-fine control for probing high density SMD circuits. Rating: 600V, CAT III, 1A.
ITEM(S) ON SALE
$21.59USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Pomona Electronics 72923-0 Mini Test Probeblck With Sharp Spring Mounted Tip EW-20009-68 Mfr # 72923-0
COMPARE
Mini Test Probeblck With Sharp Spring Mounted Tip
$21.59USD / Each
$23.99USD / Each