This product is now discontinued, View Related Products

Potassium Bisulfate

  • Acidity (as H2SO4) - 35-37%
  • Insoluble Matter and NH4OH Precipitate - ≤0.01%
  • Chloride - ≤0.001%
  • Nitrate - ≤0.005%
  • Phosphate - ≤0.001%
  • Calcium and Magnesium Precipitate - ≤0.010%
  • Heavy Metals (as Pb) - ≤0.001%
  • Iron - ≤0.002%
  • Sodium - ≤0.02%
  • Arsenic - ≤5ppm
  • Bulk Density - ≥1.17g/cc
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical P193500 Potassium Bisulfate (Cryst./Certified) (500g) EW-88078-42 Mfr # P193500
COMPARE
7646-93-7
Call For Price
Acros Organics AC207755000 Potassium bisulfate c.p. (500g) EW-88222-51 Mfr # AC207755000
COMPARE
7646-93-7
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat