This product is now discontinued, View Related Products

Ricca Chemical Potassium Hydroxide, 0.100 Normal (N/10) in Methanol

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Ricca Chemical Company 6260-32 Potassium Hydroxide, 0.100 Normal (N/10) in Methanol (1 L) EW-86981-78 Mfr # 6260-32
COMPARE
Includes 67-56-1 and 1310-58-3 1000 Potassium Hydroxide, 0.100 Normal (N/10) in Methanol (1 L)
Call For Price
Ricca Chemical Company 6260-1 Potassium Hydroxide, 0.100 Normal (N/10) in Methanol (1 gal) EW-86986-96 Mfr # 6260-1
COMPARE
Includes 67-56-1 and 1310-58-3 3785 Potassium Hydroxide, 0.100 Normal (N/10) in Methanol (1 gal)
Call For Price
Ricca Chemical Company 6260-5 Potassium Hydroxide, 0.100 Normal (N/10) in Methanol (20 L) EW-86993-85 Mfr # 6260-5
COMPARE
Includes 67-56-1 and 1310-58-3 20000 Potassium Hydroxide, 0.100 Normal (N/10) in Methanol (20 L)
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat