Potassium Permanganate Solution 23417

$141.00 - $213.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Fisher Chemical SP2884 Potassium Permanganate Solution (0.1N (N/10)/Certified) (4 liter) EW-88020-92 Mfr # SP2884
COMPARE
7722-64-7 4000 Potassium Permanganate Solution (0.1N (N/10)/Certified) (4 liter)
$208.00USD / Each
Fisher Chemical SP2821 Potassium Permanganate Solution (1N/Certified) (1 liter) EW-88085-05 Mfr # SP2821
COMPARE
7722-64-7 and 7732-18-5 1000 Potassium Permanganate Solution (1N/Certified) (1 liter)
$141.00USD / Each
Fisher Chemical SP2824 Potassium Permanganate Solution (1N/Certified) (4 liter) EW-88085-06 Mfr # SP2824
COMPARE
7722-64-7 4000 Potassium Permanganate Solution (1N/Certified) (4 liter)
$213.00USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat