Dickson Circular Pressure Chart Recorders
NEMA 4X, water-resistant enclosure withstands harsh outdoor environments
  • Keep a record of your pressure up to 1000 psi
  • Compact, lightweight design
  • Battery operated for easy portability
The PW4 weather-resistant pressure recorder is housed in a durable NEMA 4X case. These lightweight 4" or 8" recorders provide protection against corrosion, windblown dust and rain, splashing water and hose-directed water. Note: PW4 & PW8 Pressure Chart Recorders are not suitable for monitoring ammonia, acids or other corrosive substances.
$947.00 - $1,220.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chart Diameter (in) Chart Speed Max Input (VDC)
Availability Pricing
Dickson PW861 8'' Pressure chart recorder, 0 to 100 PSI, 24-hour chart EW-68511-42 Mfr # PW861
COMPARE
8 24 hour
$947.00USD / Each
Dickson PW865 8'' Pressure chart recorder, 0 to 200 PSI, 24-hour chart EW-68511-43 Mfr # PW865
COMPARE
8 24 hour
$947.00USD / Each
Dickson PW867 8'' Pressure chart recorder, 0 to 300 PSI, 24-hour chart EW-68511-44 Mfr # PW867
COMPARE
8 24 hour
$947.00USD / Each
Dickson PW875 8'' Pressure chart recorder, 0 to 1000 PSI, 24-hour chart EW-68511-45 Mfr # PW875
COMPARE
8 24 hour
$1,220.00USD / Each
Dickson PW860 8'' Pressure chart recorder, 0 to 100 PSI, 7-day chart EW-68511-46 Mfr # PW860
COMPARE
8 7 day
$947.00USD / Each
Dickson PW864 8'' Pressure chart recorder, 0 to 200 PSI, 7-day chart EW-68511-47 Mfr # PW864
COMPARE
8 7 day
$947.00USD / Each
Dickson PW866 8'' Pressure chart recorder, 0 to 300 PSI, 7-day chart EW-68511-48 Mfr # PW866
COMPARE
8 7 day
$947.00USD / Each
Showing 1- 7 of 7