Setra AccuSense™ High-Accuracy Pressure Transducer Accessories
Optimizer performance of your pressure transducer
These accessories are compatible with the Setra 98075-XX series AccuSense™ High-Accuracy Pressure Transducers.
The SecureCal™ calibrator 98075-95 is used to calibrate transducers in the field—enables zero-tare and snap-span adjustment, factory calibration restore function, and two-line LCD/LED feedback.
ITEM(S) ON SALE
$46.75 - $284.75USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Setra SRAN Remote Annunciator for Visual and Audible Status Indication EW-98074-90 Mfr # SRAN
COMPARE
Remote Annunciator for Visual and Audible Status Indication
$284.75USD / Each
$335.00USD / Each
COMPARE
Pressure Snubber for Constant Pressure Measurement
$46.75USD / Each
$55.00USD / Each
Setra 600751 Bayonet Electrical Connector Adapter, female 6-pin, ASM EW-98075-90 Mfr # 600751
COMPARE
Bayonet Electrical Connector Adapter, female 6-pin, ASM
$85.00USD / Each
$100.00USD / Each