PrimeLock® High-Purity PFA Fittings, Caps
Ultra-clean and reliable fittings for highly critical and corrosive chemicals
  • Broad chemical compatibility with PFA wetted components Three nonwetted backup seals prevent leaks without introducing sources of entrapment or flow interruptions Audible and visual checks guarantee proper connections Heat not required for fitting installation
PFA wetted fittings handle critical chemical applications for manufacturers and laboratories. Caps close and protect the unused tube end on PrimeLock fittings.
$14.10 - $34.50USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in)
Availability Pricing
COMPARE
3/4
$19.40USD / Each
COMPARE
1
$20.75USD / Each
COMPARE
1 1/4
$34.50USD / Each
COMPARE
1/4
$14.20USD / Each
COMPARE
3/8
$14.10USD / Each
COMPARE
1/2
$17.20USD / Each