PrimeLock® Hot Air Mandrels for High-Purity PFA Fittings
Easily install your High-Purity PFA Fittings
  • Optimize performance of your PFA PrimeLock® Fittings
Hot air gun mandrels are useful when space restrictions prevent the use of the handheld or bench top installation tools, sizes 1/2", 3/4" and 1".
$117.00 - $343.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With Description
Availability Pricing
COMPARE
PFA PrimeLock High-Purity PFA Flare Fittings, 1.25" OD Tubing Installation Mandrel, hot air, 1.25" fittings; 1/ea
$117.00USD / Each
COMPARE
PFA PrimeLock High-Purity PFA Flare Fittings, 1" OD Tubing Installation Mandrel, hot air, 1" fittings; 1/ea
$117.00USD / Each
COMPARE
PFA PrimeLock High-Purity PFA Flare Fittings, 1/2 to 1" OD Tubing Installation Mandrel Kit, hot air, 1/2"-1" fittings
$343.00USD / Each
COMPARE
PFA PrimeLock High-Purity PFA Flare Fittings, 3/4" OD Tubing Installation Mandrel, hot air, 3/4" fittings; 1/ea
$117.00USD / Each
COMPARE
PFA PrimeLock High-Purity PFA Flare Fittings, 1/2" OD Tubing Installation Mandrel, hot air, 1/2" fittings; 1/ea
$117.00USD / Each