PRO Scientific MaX Homogenizing Packages
Great for all types of samples, even tough or stringy
  • Included saw-tooth generator probe provides high shearing for complete sample breakdown
  • 316 stainless steel and PTFE probe construction allows sterilization with any method
  • Ideal for 5- to 50-mL tubes and small to large beakers
Use these homogenizers for a wide range of lab applications such as emulsion and suspension, cell disruption, and protein extraction. Homogenizers feature a separate ON/OFF switch allowing for the slide switch speed control to remain at your ideal homogenizing speed. Quickly and efficiently break down samples using the included saw-tooth probe(s). All probes are precision crafted from 316 SS and PTFE to be chemically compatible with your sample.
$2,635.00 - $5,155.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Generator Included Power (VAC)
Availability Pricing
PRO Scientific Standard MaX-Homogenizer Package, 10 to 50 mL Tubes; 120V EW-04751-01 Mfr # PK-01200MXS
COMPARE
10-mm dia saw-tooth 120
$2,635.00USD / Each
PRO Scientific Deluxe MaX-Homogenizer Package, 10 mL Tubes to 2 L Beaker; 120V EW-04751-11 Mfr # PK-01250MXD
COMPARE
10- and 20-mm saw-tooth 120
$4,785.00USD / Each
PRO Scientific Premium MaX-Homogenizer Package, 10 mL Tubes to 2 L Beaker; 120V EW-04751-21 Mfr # PK-01250MXP
COMPARE
10- and 20-mm saw-tooth 120
$5,155.00USD / Each