Propylene Carbonate

  • Assay (GC) - ≥98%
$67.00 - $331.00USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Acros Organics AC131560025 Propylene Carbonate, 99.5%, 2.5 L EW-88134-57 Mfr # AC131560025
COMPARE
108-32-7 99.5% 2500
$331.00USD / Each
Acros Organics AC131565000 Propylene Carbonate, 99.5%, 500 mL EW-88134-59 Mfr # AC131565000
COMPARE
108-32-7 99.5% 500
$67.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat