This product is now discontinued, View Related Products

Propylene Oxide

  • Assay - ≥98%
  • Color (APHA) - ≤10
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical O43321 Propylene Oxide (Reagent) (1 liter) EW-88077-98 Mfr # O43321
COMPARE
75-56-9
Call For Price
Acros Organics AC149620010 Propylene oxide 99% (1l) EW-88150-35 Mfr # AC149620010
COMPARE
75-56-9
Call For Price
Acros Organics AC149620025 Propylene oxide 99% (2.5l) EW-88150-36 Mfr # AC149620025
COMPARE
75-56-9
Call For Price
Acros Organics AC149620500 Propylene oxide 99% (50ml) EW-88150-38 Mfr # AC149620500
COMPARE
75-56-9
Call For Price
Acros Organics AC220160010 Propylene oxide p.a. (1l) CAS 75-56-9 (1,2-Epoxypropane, Methyloxirane) EW-88235-47 Mfr # AC220160010
COMPARE
75-56-9 Pro Analysi
Call For Price
Acros Organics AC220160050 Propylene oxide p.a. (5ml) EW-88235-48 Mfr # AC220160050
COMPARE
75-56-9
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat