ProtEX-MAX Explosion-Proof Instruments
Die-cast aluminum NEMA 7 & 4X enclosures with SafeTouch<@> through glass buttons
  • View from wide angles
  • Display mountable at 0°, 90°, 180°. and 270°
  • Easy field setup and wiring
  • RS-485 with Modbus protocol standard
  • Four 3/4" NPT conduit holes for wiring
  • Flanges for wall or pipe mounting
ProtEX-MAX series offers a wide range of explosion-proof process meters, flow rate/totalizers, batch controllers, termperature meters, dual-input process meters, and Modbus<@> meters with standard dual line, or large 1.2" displays. Unique SafeTouch<@> through glass buttons allow for in-place programming.
$1,930.00 - $2,360.00USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Digits Display Type Description
Availability Pricing
Precision Digital PD8-6000-6H0 Dual-line process meter EW-30004-70 Mfr # PD8-6000-6H0
COMPARE
6 0.6" LED; 0.46" LED Dual-line process meter
$2,000.00USD / Each
Precision Digital PD8-6000-6H5 Dual-line process meter EW-30004-71 Mfr # PD8-6000-6H5
COMPARE
6 0.6" LED; 0.46" LED Dual-line process meter
$2,040.00USD / Each
Precision Digital PD8-6000-6H7 Dual-line process meter EW-30004-72 Mfr # PD8-6000-6H7
COMPARE
6 0.6" LED; 0.46" LED Dual-line process meter
$2,190.00USD / Each
Precision Digital PD8-6060-6H0 Dual-line, Dual-Input process meter EW-30004-73 Mfr # PD8-6060-6H0
COMPARE
6 0.6" LED; 0.46" LED Dual-line, Dual-Input process meter
$2,150.00USD / Each
Precision Digital PD8-6060-6H5 Dual-line, Dual-Input process meter EW-30004-74 Mfr # PD8-6060-6H5
COMPARE
6 0.6" LED; 0.46" LED Dual-line, Dual-Input process meter
$2,190.00USD / Each
Precision Digital PD8-6060-6H7 Dual-line, Dual-Input process meter EW-30004-76 Mfr # PD8-6060-6H7
COMPARE
6 0.6" LED; 0.46" LED Dual-line, Dual-Input process meter
$2,340.00USD / Each
Precision Digital PD8-6200-6H0 Explosion-Proof Flow Rate/Totalizer EW-30004-77 Mfr # PD8-6200-6H0
COMPARE
6 0.6" LED; 0.46" LED Explosion-Proof Flow Rate/Totalizer
$2,040.00USD / Each
Precision Digital PD8-6200-6H5 Explosion-Proof Flow Rate/Totalizer; 2 Relay EW-30004-78 Mfr # PD8-6200-6H5
COMPARE
6 0.6" LED; 0.46" LED Explosion-Proof Flow Rate/Totalizer; 2 Relay
$2,080.00USD / Each
Precision Digital PD8-6200-6H7 Explosion-Proof Flow Rate/Totalizer; 4 Relay EW-30004-79 Mfr # PD8-6200-6H7
COMPARE
6 0.6" LED; 0.46" LED Explosion-Proof Flow Rate/Totalizer; 4 Relay
$2,230.00USD / Each
Precision Digital PD8-6210-6H5 Explosion-Proof Batch Controller; 2 Relay EW-30004-80 Mfr # PD8-6210-6H5
COMPARE
6 0.6" LED; 0.46" LED Explosion-Proof Batch Controller; 2 Relay
$2,200.00USD / Each
Precision Digital PD8-6210-6H7 Explosion-Proof Batch Controller; 4 Relay EW-30004-88 Mfr # PD8-6210-6H7
COMPARE
6 0.6" LED; 0.46" LED Explosion-Proof Batch Controller; 4 Relay
$2,360.00USD / Each
Precision Digital PD8-765-6X0-10 Explosion-Proof Process/Temp Meter EW-30004-89 Mfr # PD8-765-6X0-10
COMPARE
4 1.2" LED Explosion-Proof Process/Temp Meter
$1,950.00USD / Each
Precision Digital PD8-765-6X2-10 Explosion-Proof Process/Temp Meter; 2 Relay EW-30004-90 Mfr # PD8-765-6X2-10
COMPARE
4 1.2" LED Explosion-Proof Process/Temp Meter; 2 Relay
$1,930.00USD / Each
Precision Digital PD8-765-6X3-20 Explosion-Proof Process/Temp Meter; 4-20mA EW-30004-91 Mfr # PD8-765-6X3-20
COMPARE
4 1.2" LED Explosion-Proof Process/Temp Meter; 4-20mA
$2,150.00USD / Each
Showing 1- 14 of 14
Need Help with this product?
Call Us At1-847-549-7600
Live chat