ProtEX-RTs Explosion-Proof Rate/Totalizers

Program your process meter on-site
  • 5-digit rate and 7-character total/tag display
  • Backlight standard
  • Rotatable display
  • FM, CSA, ATEX, IECEx, CE approved
The ProtEX-RTs are rugged, explosion-proof, full-featured rate/totalizers designed for demanding applications in hazardous areas or in the harshest environmental conditions. The PD6820 accepts analog inputs and the PD6830 accepts pulse inputs. They are programmed using the four SafeTouch™ through-glass buttons without removing the cover, or the four mechanical buttons. The sleek die-cast aluminum enclosure is provided with two ¾-inch threaded conduit holes and integrated pipe or wall mounting flanges.
$859.00 - $999.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Display Digits Display Type Enclosure
Availability Pricing
Precision Digital Explosion-Proof Rate/Totalizer with battery, backlight, and loop output. EW-30004-62 Mfr # PD6830-BTA-0
COMPARE
5; 7 0.7" LED; 0.4" LED NEMA 4X, 7, 9; IP68; explosion-proof die-cast aluminum
$999.00USD / Each
Precision Digital Explosion-Proof Rate/Totalizer, 9-30 VDC, with backlight. EW-30004-63 Mfr # PD6830-AP0-0
COMPARE
5; 7 0.7" LED; 0.4" LED NEMA 4X, 7, 9; IP68; explosion-proof die-cast aluminum
$859.00USD / Each
Precision Digital PD6820-0K1 Explosion-Proof Rate/Totalizer, Input Loop Powered EW-30004-66 Mfr # PD6820-0K1
COMPARE
5; 7 0.7" LED; 0.4" LED NEMA 4X, 7, 9; IP68; explosion-proof die-cast aluminum
$859.00USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat