This product is now discontinued, View Related Products
Protractors
Complete 360° angle measurements
Bevel Protractor features a vernier dial with 5° graduations. Attached magnifier offers easier viewing of dial. Protractor includes 6” and 12” rulers, and an angle attachment finished in satin chrome, and a fitted case.
Universal Dial Protractor features a wide spaced dial with graduations of 5°. Protractor includes 6” and 12” rulers, and lock and acute angle attachments made of hardened stainless steel with a satin chrome finish. Fitted case also included.
Digital Protractor features a digital LCD for inclination values. Measurements are generated by an electronic gravity sensor and processed through an electronic circuit. Other features include alternate zero (reference), display hold, and a machined aluminum frame.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max range (in) Resolution
Availability Pricing
Fowler 52-440-612 Bevel Protractor, Satin Chrome EW-09918-70 Mfr # 52-440-612
COMPARE
Call For Price
Fowler 52-440-777 Universal dial protractor EW-09918-72 Mfr # 52-440-777
COMPARE
Call For Price
Fowler 54-950-315 Pro-360 Digital Protractor, 360° range EW-97241-60 Mfr # 54-950-317
COMPARE
0.1°