PTFE ACR Valves for Chemical and Corrosive Fluids
Chemical-resistant valves provide precise fluid control
  • Manufactured in a clean room environment
  • Process connections of NPT(F), compression, and hose barb
  • Long service life performing in the harshest of applications
These pneumatically operated valves feature chemically resistant wetted surfaces constructed from PTFE—the material of choice for both thermal resistance and chemically inert applications, or many difficult-to-handle media. Commonly used in wet benches and process work stations, these valves are ideal for DI water and other harsh applications. Valves are ideal for flow control of high purity or corrosive chemicals, and feature one common inlet pressure port and two discreet outlet ports that are activated via pneumatic trigger signal.
Valves are an ideal solution for flow control in the research, biological, food, and chemical processing, as well as semiconductor manufacturing and petrochemical environments.
$448.00 - $685.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type Inlet Port Orifice (in)
Availability Pricing
COMPARE
NPT(F) 1/8" NPT(F) 0.125
$460.00USD / Each
COMPARE
NPT(F) 1/4" NPT(F) 0.25
$460.00USD / Each
COMPARE
NPT(F) 3/8" NPT(F) 0.375
$523.00USD / Each
COMPARE
NPT(F) 1/2" NPT(F) 0.5
$685.00USD / Each
COMPARE
Compression 1/8" compression 0.125
$448.00USD / Each
COMPARE
Compression 1/4" compression 0.25
$448.00USD / Each
COMPARE
Compression 3/8" compression 0.375
$523.00USD / Each
COMPARE
Compression 1/2" compression 0.5
$553.00USD / Each
COMPARE
Flare 1/8" Flare 0.125
$495.00USD / Each
COMPARE
Barbed 1/4" Barb 0.25
$510.00USD / Each
COMPARE
Barbed 3/8" Barb 0.375
$648.00USD / Each
COMPARE
Barbed 1/2" Barb 0.5
$648.00USD / Each
Showing 1- 12 of 12
Need Help with this product?
Call Us At1-847-549-7600
Live chat