This product is now discontinued, View Related Products
PTFE Pressure Regulators for Liquids
Pressure regulators are designed for use in high purity water and aggressive chemicals applications. Designed with a PTFE wetted flow path, the unit is ideal for DI water systems and corrosive media found in solar, semiconductor, pharmaceutical and chemical process applications.
All configurations are fitted for panel mounting with an easy-to-grip adjustment knob. PTFE coated stainless steel adjustment screw is configured for low torque, reliable manual operation.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Max Pressure (PSI) Orifice (in)
Availability Pricing
COMPARE
1/4\" Flare 0.1875
Call For Price
COMPARE
1/4\" Flare 0.1875
Call For Price
COMPARE
1/4\" NPT(F) 0.25
Call For Price
COMPARE
1/4\" NPT(F) 0.25