PTFE Sampling Metering Valve
Sample valve is designed for use in high purity water and aggressive chemical applications. Designed with a PTFE wetted flow path, the unit is ideal for DI Water Systems and corrosive media found in solar, semiconductor, pharmaceutical and chemical process applications. Valve is panel-mount and functions with a manual push level to actuate a metered flow, ensuring a controlled dispensing of fluid for manual sampling.
Commonly used for dispensing controlled amounts of media for element verification tests, manual dispensing of DI Water into carboy and other transport containers, and for dispensing metered amounts of chemicals to blend and balance process chemistries.
$388.00 - $428.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Orifice (in) Outlet Port
Availability Pricing
COMPARE
1/4" Flare 0.125 1/4" Flare
$428.00USD / Each
COMPARE
1/4" NPT(F) 0.250 1/4" NPT(F)
$388.00USD / Each
COMPARE
1/4" compression 0.188 1/4" compression
$408.00USD / Each