Pulsafeeder Economical Peristaltic Pumps
Easy maintenance and no contamination—tubing is the only wetted part
  • Easy tube change-out minimizes downtime between tubing set changes
  • Inherent degassing function and rugged gear train offer many years of reliable service
These simple-to-use, self-priming peristaltic pumps deliver worry-free dosing.
$461.00 - $812.00USD / Each
31 variations of this product are available.
See All Product Options Below
31 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Pressure (PSI) Power (VAC)
Availability Pricing
Pulsafeeder XP004LFNX Pump, Fixed Peristaltic Pump, 4 GPD (0.6 LPH), 100 PSI (6.8 BAR), 115 VAC, 60Hz EW-74130-11 Mfr # XP004LFHX
COMPARE
10 100 115
$469.00USD / Each
Pulsafeeder XP023LFNX Fixed Speed Peristaltic Pump, 3.6 LPH, 100 PSI (6.8 BAR), 115 VAC EW-74130-16 Mfr # XP023LFHX
COMPARE
3600 6.8 115
$464.00USD / Each
Pulsafeeder XP030LFNX Fixed-Speed Peristaltic Pump; 30GPD/80PSI/115V/60Hz EW-74130-17 Mfr # XP030LFHX
COMPARE
4700 80 115
$464.00USD / Each
Pulsafeeder XP050LFLX Fixed-Speed Peristaltic Pump; 50GPD/80PSI/115V/60Hz EW-74130-18 Mfr # XP050LFLX
COMPARE
7900 80 115
$469.00USD / Each
Pulsafeeder XP055LVNX Fixed Speed Peristaltic Pump, 8.67 LPH, 80 PSI (5.4 BAR), 115 VAC EW-74130-24 Mfr # 7413024
COMPARE
145 80 115
$812.00USD / Each
Pulsafeeder XP0075ANX Adjustable-Speed Peristaltic Pump; 7GPD/100PSI/230V/50Hz EW-74130-72 Mfr # XP0075ANX
COMPARE
18 100 230
$635.00USD / Each
Pulsafeeder XP0095ANX Adjustable-Speed Peristaltic Pump; 9GPD/100PSI/230V/50Hz EW-74130-73 Mfr # XP0095ANX
COMPARE
23 100 230
$635.00USD / Each
Pulsafeeder XP0155ANX Adjustable-Speed Peristaltic Pump; 15GPD/100PSI/230V/50Hz EW-74130-74 Mfr # XP0155ANX
COMPARE
40 100 230
$635.00USD / Each
Pulsafeeder XP0305ANX Adjustable-Speed Peristaltic Pump; 30GPD/80PSI/230V/50Hz EW-74130-77 Mfr # XP0305ANX
COMPARE
78 80 230
$635.00USD / Each
Pulsafeeder XP0505ANX Adjustable-Speed Peristaltic Pump; 50GPD/80PSI/230V/50Hz EW-74130-78 Mfr # XP055LVHX
COMPARE
132 80 230
$807.00USD / Each
Pulsafeeder XP0805ANX Adjustable-Speed Peristaltic Pump; 80GPD/25PSI/230V/50Hz EW-74130-79 Mfr # XP0805ANX
COMPARE
210 25 230
$635.00USD / Each
Pulsafeeder XP004HFNX Pump, Fixed Peristaltic Pump, 4 GPD (0.6 LPH), 100 PSI (6.8 BAR), 230 VAC, 60Hz EW-74130-81 Mfr # XP004HFHX
COMPARE
10 100 230
$645.00USD / Each
Pulsafeeder XP007HFNX Pump, Fixed Peristaltic Pump, 7 GPD (1.1 LPH), 100 PSI (6.8 BAR), 230 VAC, 60Hz EW-74130-82 Mfr # XP007HFHX
COMPARE
18 100 230
$464.00USD / Each
Pulsafeeder XP009HFNX Pump, Fixed Peristaltic Pump, 9 GPD (1.4 LPH), 100 PSI (6.8 BAR), 230 VAC, 60Hz EW-74130-83 Mfr # XP009HFHX
COMPARE
23 100 230
$464.00USD / Each
Pulsafeeder XP030HFNX Fixed-Speed Peristaltic Pump; 30GPD/80PSI/230V/60Hz EW-74130-87 Mfr # XP030HFHX
COMPARE
78 80 230
$464.00USD / Each
Pulsafeeder XP050HFNX Fixed-Speed Peristaltic Pump; 50GPD/80PSI/230V/60Hz EW-74130-88 Mfr # XP050HFHX
COMPARE
132 80 230
$469.00USD / Each
Pulsafeeder XP080HFNX Fixed Speed Peristaltic Pump, 80 GPD (12.6 LPH), 25 PSI (1.7 BAR), 230 VAC, 60Hz EW-74130-89 Mfr # XP080HFLX
COMPARE
210 25 230
$469.00USD / Each
Pulsafeeder XP008HVNX Variable Speed Peristaltic Pumps, 1.26 LPH, 100 PSI, 230VAC EW-74130-91 Mfr # XP008HVHX
COMPARE
21 100 230
$812.00USD / Each
Pulsafeeder XP033HVNX Variable Speed Peristaltic Pump, 33 GPD (5.2 LPH), 100 PSI, 230 VAC EW-74130-93 Mfr # XP033HVHX
COMPARE
87 100 230
$812.00USD / Each
Pulsafeeder XP055HVNX Variable Speed Peristaltic Pump, 55 GPD (8.7 LPH), 80 PSI, 230 VAC EW-74130-94 Mfr # XP055HVHX
COMPARE
145 80 230
$812.00USD / Each
Showing 1 - 20 of 31