Pulsafeeder Manual Control Metering Pumps
Reliable metering performance – guided dual check valves provide precise seating and excellent priming and suction lift
  • PVC and PTFE construction with external control
  • Ideal for water treatment and chemical feed applications
Manual control pumps feature adjustable stroke rate and length, reliable timing circuits, and circuit protection from voltage and current upsets. Leak-free bleed valves provide safe and easy priming. Pumps are water resistant and can be installed outdoors.
$475.00 - $2,018.00USD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPH) Max Flow Rate (mL/min) Power (VAC)
Availability Pricing
Pulsafeeder LE44SA-VTC1 Manual Control Metering Pump, 44 gdp, 115 VAC EW-74142-00 Mfr # LE44SA-VTC1
COMPARE
1.83 116 115
$794.00USD / Each
Pulsafeeder LE44SB-VTC1 Manual Control Metering Pump, 44 gdp, 230 VAC EW-74142-05 Mfr # LE44SB-VTC1
COMPARE
1.83 116 230
$827.00USD / Each
Pulsafeeder LEG5SA-VTC3 Manual Control Metering Pump, 94 gdp, 115 VAC EW-74142-10 Mfr # LPG5SA-VTC3-XXX
COMPARE
3.92 247 115
$1,221.00USD / Each
Pulsafeeder LEG5SB-VTC3-XXX Manual Control Metering Pump, 94 gdp, 230 VAC EW-74142-15 Mfr # LPG5SB-VTC3-XXX
COMPARE
3.92 247 230
$1,243.00USD / Each
Pulsafeeder LEH7SA-WTC3 Manual Control Metering Pump, 240 gdp, 115 VAC EW-74142-20 Mfr # LPH7SA-WTC3-XXX
COMPARE
10 631 115
$1,380.00USD / Each
Pulsafeeder LEH7SB-WTC3 Manual Control Metering Pump, 240 gdp, 230 VAC EW-74142-25 Mfr # LPH7SB-WTC3-XXX
COMPARE
10 631 230
$1,413.00USD / Each
Pulsafeeder LEH8SA-VTCB Manual Control Metering Pump, 500 gdp, 115 VAC EW-74142-30 Mfr # LPH8SA-WTCB-XXX
COMPARE
20.83 1314 115
$1,974.00USD / Each
Pulsafeeder LEH8SB-VTCB Manual Control Metering Pump, 500 gdp, 230 VAC EW-74142-35 Mfr # LPH8SB-WTCB-XXX
COMPARE
20.83 1314 230
$2,018.00USD / Each
Pulsafeeder LD02SA-VTC1-XXX Manual Control Metering Pump, 6 gdp, 115 VAC EW-78118-00 Mfr # LD02SA-VTC1-XXX
COMPARE
0.25 16 115
$478.00USD / Each
Pulsafeeder LD02SB-VTC1-XXX Manual Control Metering Pump, 6 gdp, 230 VAC EW-78118-05 Mfr # LD02SB-VTC1-XXX
COMPARE
0.25 16 230
$502.00USD / Each
Pulsafeeder LD03SA-VTC1-XXX Manual Control Metering Pump, 12 gdp, 115 VAC EW-78118-10 Mfr # LD03SA-VTC1-XXX
COMPARE
0.5 32 115
$478.00USD / Each
Pulsafeeder LD03SB-VTC1-XXX Manual Control Metering Pump, 12 gdp, 230 VAC EW-78118-15 Mfr # LD03SB-VTC1-XXX
COMPARE
0.5 32 230
$512.00USD / Each
Pulsafeeder LD04SA-VTC1-XXX Manual Control Metering Pump, 22 gdp, 115 VAC EW-78118-20 Mfr # LD04SA-VTC1-XXX
COMPARE
0.92 58 115
$478.00USD / Each
Pulsafeeder LD04SB-VTC1-XXX Manual Control Metering Pump, 22 gdp, 230 VAC EW-78118-25 Mfr # LD04SB-VTC1-XXX
COMPARE
0.92 58 230
$502.00USD / Each
Pulsafeeder LD54SA-VTC1-XXX Manual Control Metering Pump, 30 gdp, 115 VAC EW-78118-30 Mfr # LD54SA-VTC1-XXX
COMPARE
1.25 79 115
$475.00USD / Each
Pulsafeeder LD54SB-VTC1-XXX Manual Control Metering Pump, 30 gdp, 230 VAC EW-78118-35 Mfr # LD54SB-VTC1-XXX
COMPARE
1.25 79 230
$502.00USD / Each
Showing 1- 16 of 16