Pulsafeeder Mechanical Diaphragm Pumps
Heavy duty – get up to 150 psi with maximum flow rates up to 140 GPH
  • Mechanical diaphragm design eliminates leakage and contamination of fluid
  • Stroke lock maintains ±2% accuracy of the stroke setting
  • Lubricated for life — sealed cast iron housing reduces wear and is rated NEMA 4, IP55
Manual control and automatic control both utilize 1/2 hp, TEFC motor which provides for a quiet and cool operation. This lightweight metering pump allows a 1000:1 turn down capability by using the MPC controller. The MPC automatic controller controls the pump speed and flow through a digital display. The MPC uses digital (4 to 20 mA) inputs and outputs to monitor and control elements of the process.
The PVC model is ideal for pumping Sodium Hydroxide, Sulfuric acids in low concentrations, and Hyrdrochloric acids in low concentrations. Choose the stainless steel version for moving Acetic acids, and Lithium Cholrides.
$776.00 - $854.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Max Pressure (PSI) Wetted Materials
Availability Pricing
Pulsafeeder NLK040AP OMNI Service Kit for 30 and 65 gph pumps, 316 SS liquid end EW-74148-93 Mfr # NLK040AP
COMPARE
$776.00USD / Each
Pulsafeeder NLK050AP OMNI Service Kit for 108 gph pumps, 316 SS liquid end EW-74148-95 Mfr # NLK050AP
COMPARE
$854.00USD / Each