Pump and Mix Tube Sets
These new tube sets eliminate the hassle of needing a separate mixing tube. With the unique lever design, you can easily switch between pumping and mixing modes to pump and mix the drum contents separately or simultaneously. SS tube sets are 19/16" OD with 1" hose barb; PP tube sets are 2" OD with 3/4", 1", or 1-1/4" hose barb.
$1,136.50 - $2,239.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Tube Length, Inlet (in) Wetted Materials
Availability Pricing
Flux F426S 41/38 39" Pump and mix tube set, 39", Stainless Steel EW-07076-50 Mfr # F426S 41/38 39"
COMPARE
45 39 Stainless steel
$1,493.00USD / Each
Flux F426S 41/38 60" Pump and mix tube set, 60", Stainless Steel EW-07076-54 Mfr # F426S 41/38 60"
COMPARE
45 60 Stainless steel
$2,239.00USD / Each
Flux F426P 50/33 47" Pump and mix tube set, 47", Polypropylene EW-07076-58 Mfr # F426P 50/33 47"
COMPARE
45 47 Polypropylene
$1,136.50USD / Each