Pura-Lite 120 VAC Resistivity Indicators

A low-cost alternative to digital purity meters
  • Designed for use with B-Pure systems
These indicators can be mounted directly on top of single- or doublecartridge housings with 1⁄2" NPT(F) connections.
$237.75 - $314.00USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description Resistivity Threshold
Availability Pricing
Thermo Scientific Barnstead E3450 Pura -Lite Resistivity Indicators, 50 k-ohm, 120 VAC EW-01504-36 Mfr # E3450
COMPARE
Pura -Lite Resistivity Indicators, 50 k-ohm, 120 VAC 50 kΩ
$314.00USD / Each
COMPARE
-Lite Resistivity Indicators, 1 m-ohm, 120 VAC 200 kΩ
$237.75USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat