This product is now discontinued, View Related Products
Push-Button Temperature Transmitters (RTD, Thermocouple, Thermistor)
Customize your temperature range with a button press
  • LED indicates programming operations and out-of-range sensor
  • Drift-free digital linearization
  • Fits standard DIN probe head enclosures
These economical transmitters feature a push-button, customizable temperature range. Simply connect a power supply and a simulator/calibrator to the transmitter, select the desired temperature at 4 or 20 mA, and hold the set button until the LED indicator blinks. Transmitters are factory set at a range of 32 to 212°F (0 to 100°C).
Thermocouple Transmitter is selectable for Type J, K, or T probes. It features galvanic isolation.
RTD Transmitter connects to any standard 3-wire Pt 100 RTD temperature sensor. The RTD model also allows push-button calibration.
Thermistor Transmitter is compatible with 400 series thermistor probes for excellent accuracy over the ambient temperature range.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Loop Power Supply
Availability Pricing
COMPARE
Call For Price
COMPARE
13 to 40 VDC
Call For Price
COMPARE
13 to 40 VDC