PVC Reinforced Tubing
Applications: High-pressure use, food processing, and beverage handling.
Characteristics: Good acid/alkali resistance and aging characteristics. White polyester yarn with orange tracer yarn embedded in a transparent PVC wall.
Certification: Resin is FDA and NSF-51 compliant.
Temperature range: -50 to 175°F (-45.5 to 79.4°C). NSF temperature to 140°F.
Sterilize: Ethylene oxide.
$137.00 - $1,401.00USD / Pkg
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
100 3/16 3/8
$137.00USD / Pkg
COMPARE
100 3/8 9/16
$177.00USD / Pkg of 1
COMPARE
100 1/4 7/16
$162.00USD / Pkg
COMPARE
100 3/8 9/16
$198.00USD / Pkg
COMPARE
100 1/2 3/4
$221.00USD / Pkg
COMPARE
100 5/8 29/32
$286.00USD / Pkg
COMPARE
100 3/4 1 1/32
$304.00USD / Pkg
COMPARE
100 1 1 5/16
$370.00USD / Pkg
COMPARE
100 1 1/2 1 5/16
$791.00USD / Pkg
COMPARE
100 2 2 1/2
$1,401.00USD / Pkg
Showing 1- 10 of 10