Pyrex Brand 1595 Bottles
These bottles are ideal for solution storage and media preparation. The 19-liter size is weight controlled to 15-lb maximum for ease of handling. Bottle necks are tooled to increase mechanical strength and ensure uniform stopper fit. Caution: Thick-walled glassware such as jars, bottles, cylinders an filter flasks, should never be heated on a hot plate, as this greatly increases the chance of breakage.
$421.00 - $901.00USD / Case of 4
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
Pyrex 1595-2X Brand 1595 Solution Bottle; conventional bottle shape, 9.5 L
Buy More Save More
EW-34542-21 Mfr # 1595-2X
COMPARE
9.5 #12 Glass
$897.00USD / Case of 4
Pyrex 1595-3X Brand 1595 Solution Bottle; conventional bottle shape, 13.25 L
Buy More Save More
EW-34542-22 Mfr # 1595-3X
COMPARE
13.25 #12 Glass
$901.00USD / Case of 4
Pyrex 1595-5 Brand 1595 Solution Bottle; carboy shape, 19 L
Buy More Save More
EW-34542-23 Mfr # 1595-5
COMPARE
19 #12 Glass
$421.00USD / Case of 1
Pyrex 1595-12 Brand 1595 Solution Bottle; carboy shape, 45.5 L
Buy More Save More
EW-34542-24 Mfr # 1595-12
COMPARE
45.5 #12 Glass
$897.00USD / Case of 1