PYREX Brand 4985 Screw-Cap Erlenmeyer Flasks
  • Screw-cap
  • Graduations
These Erlenmeyer flasks with phenolic screw caps are ideal for culturing, mixing, and storing media. Impact resistant caps are equipped with cemented-in, inert white rubber liners which are resistant to steam sterilization. Flasks are graduated and include marking space.
$249.00 - $810.00USD / Pkg of 6
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Pyrex 4985-50 Brand 4985 flask with screw cap; 50 mL, pack of 12 EW-34554-01 Mfr # 4985-50
COMPARE
50 10 Glass
$465.00USD / Pkg of 12
Pyrex 4985-125 Brand 4985 flask with screw cap; 125 mL, pack of 12 EW-34554-02 Mfr # 4985-125
COMPARE
125 25 Glass
$460.00USD / Pkg of 12
Pyrex 4985-250 Brand 4985 flask with screw cap; 250 mL, pack of 6 EW-34554-03 Mfr # 4985-250
COMPARE
250 25 Glass
$249.00USD / Pkg of 6
Pyrex 4985-500 Brand 4985 flask with screw cap; 500 mL, pack of 6 EW-34554-04 Mfr # 4985-500
COMPARE
500 50 Glass
$269.00USD / Pkg of 6
Pyrex 4985-1L Brand 4985 flask with screw cap; 1000 mL, pack of 6 EW-34554-05 Mfr # 4985-1L
COMPARE
1000 50 Glass
$334.00USD / Pkg of 6
Pyrex 4985-50 Brand 4985 flask with screw cap; 50 mL, case of 24 EW-34554-21 Mfr # 4985-50
COMPARE
50 10 Glass
$758.00USD / Case of 24
Pyrex 4985-125 Brand 4985 flask with screw cap; 125 mL, case of 24 EW-34554-22 Mfr # 4985-125
COMPARE
125 25 Glass
$748.00USD / Case of 24
Pyrex 4985-250 Brand 4985 flask with screw cap; 250 mL, case of 24 EW-34554-23 Mfr # 4985-250
COMPARE
250 25 Glass
$800.00USD / Case of 24
Pyrex 4985-500 Brand 4985 flask with screw cap; 500 mL, case of 24 EW-34554-24 Mfr # 4985-500
COMPARE
500 50 Glass
$810.00USD / Case of 24
Pyrex 4985-1L Brand 4985 flask with screw cap; 1000 mL, case of 12 EW-34554-25 Mfr # 4985-1L
COMPARE
1000 50 Glass
$547.00USD / Case of 12
Showing 1- 10 of 10
Need Help with this product?
Call Us At1-847-549-7600
Live chat