Pyrex® Conical-Bottom, Disposable Glass, Centrifuge Tubes
Corning disposable glass centrifuge tubes are manufactured from PYREX® Type I borosilicate glass, which has more consistent uniform wall thickness. Tubes feature with a threaed rim for secure sealing. Caps are sold separately.
For closures, select either disposable phenolic caps with rubber liners, or reusable screw caps with PTFE liners for improved chemical resistance.
$146.00 - $152.00USD / Case of 125
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Capacity (mL) Tube Material Description
Availability Pricing
Pyrex 99502-5 Conical-Bottom Disposable Glass Centrifuge Tubes, Theaded Top, 5 mL; 125/Cs EW-34534-06 Mfr # 99502-5
COMPARE
5 Glass Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 5 ml, 125/cs
$151.00USD / Case of 125
Pyrex 99502-10 Conical-Bottom Disposable Glass Centrifuge Tubes, Theaded Top, 10 mL; 125/Cs EW-34534-08 Mfr # 99502-10
COMPARE
10 Glass Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 125/cs
$146.00USD / Case of 125
Pyrex 99502-15 Conical-Bottom Disposable Glass Centrifuge Tubes, Theaded Top, 15 mL; 125/Cs EW-34534-10 Mfr # 99502-15
COMPARE
15 Glass Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 125/cs
$152.00USD / Case of 125