PyroGuard FR Flame-Retardant Coveralls
Disposable coveralls that protect primary flame-resistant apparel
  • Flame retardant, water repellent
Coveralls are made with a tough, breathable, and lightweight material constructed of a hydro-entangled wood pulp and polyester. The surface is treated with a phosphate-based flame retardant chemical. These coveralls are intended to be worn over primary flame-resistant apparel, protecting it from dirt, grease, and non-toxic liquids. Passes NFPA 701 for vertical flammability.
$190.90 - $236.40USD / Case of 25
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Size Description
Availability Pricing
International Enviroguard Coveralls, elastic wrists, medium, 25/CS SKU 8623445 EW-86234-45 Mfr # 9012-M
COMPARE
Medium Coveralls, elastic wrists, medium, 25/CS
$190.90USD / Case of 25
International Enviroguard Coveralls, elastic wrists, large, 25/cs SKU 8623446 EW-86234-46 Mfr # 9012-L
COMPARE
Large Coveralls, elastic wrists, large, 25/cs
$190.90USD / Case of 25
International Enviroguard Coveralls, elastic wrists, extra large, 25/cs SKU 8623447 EW-86234-47 Mfr # 9012-XL
COMPARE
X-large Coveralls, elastic wrists, extra large, 25/cs
$190.90USD / Case of 25
International Enviroguard Coveralls, elastic wrists, double extra large, 25/cs SKU 8623448 EW-86234-48 Mfr # 9012-2XL
COMPARE
XX-large Coveralls, elastic wrists, double extra large, 25/cs
$190.90USD / Case of 25
International Enviroguard Coveralls, with hood, elastic wrists, medium, 25/cs SKU 8623455 EW-86234-55 Mfr # 9015-M
COMPARE
Medium Coveralls, with hood, elastic wrists, medium, 25/cs
$218.20USD / Case of 25
International Enviroguard Coveralls, with hood, elastic wrists, large, 25/cs EW-86234-56 Mfr # 9015-L
COMPARE
Large Coveralls, with hood, elastic wrists, large, 25/cs
$218.20USD / Case of 25
International Enviroguard Coveralls, with hood, elastic wrists, extra large, 25/cs SKU 8623457 EW-86234-57 Mfr # 9015-XL
COMPARE
X-large Coveralls, with hood, elastic wrists, extra large, 25/cs
$218.20USD / Case of 25
International Enviroguard Coveralls, with hood, elastic wrists, double extra large, 25/ cs EW-86234-58 Mfr # 9015-2XL
COMPARE
XX-large Coveralls, with hood, elastic wrists, double extra large, 25/ cs
$218.20USD / Case of 25
International Enviroguard Coveralls, with hood and boots, elastic wrists, medium, 25/cs SKU 8623465 EW-86234-65 Mfr # 9019-M
COMPARE
Medium Coveralls, with hood and boots, elastic wrists, medium, 25/cs
$234.40USD / Case of 25
International Enviroguard Coveralls, with hood and boots, elastic wrists, large, 25/cs SKU 8623466 EW-86234-66 Mfr # 9019-L
COMPARE
Large Coveralls, with hood and boots, elastic wrists, large, 25/cs
$234.40USD / Case of 25
International Enviroguard PyroGuard FR Flame-Retardant Coveralls, Elastic Wrist, Hood/Boots, XXL; 25/cs EW-86234-68 Mfr # 9019-2XL
COMPARE
XX-large PyroGuard FR Flame-Retardant Coveralls, Elastic Wrist, Hood/Boots, XXL; 25/cs
$236.40USD / Case of 25
Showing 1- 11 of 11