Cole-Parmer® Essentials Pre-Cleaned Square Wide-Mouth Bottles, Clear Glass
Save time—no need to prewash bottles
  • Pre-cleaned by vacuum and ionization
  • Individually sealed for long term storage with no contamination
The vacuum and ionization process removes contaminants such as dirt, dust, carton lint, oil, aerosols, and fine glass particles from these bottles. Ideal for chemical and resin sampling. Bottles come with PTFE lined caps.
$125.00 - $197.00USD / Case of 48
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
Cole-Parmer Essentials GLC-01243 Pre-Cleaned Square Wide-Mouth Bottle, Clear Glass, 15mL; 48/CS EW-34610-00 Mfr # GLC-01243
COMPARE
15 20 mm Glass
$163.00USD / Case of 48
Cole-Parmer Essentials GLC-01263 Pre-Cleaned Square Wide-Mouth Bottle, Clear Glass, 30mL; 48/CS EW-34610-10 Mfr # GLC-01263
COMPARE
30 24 mm Glass
$170.00USD / Case of 48
Cole-Parmer Essentials GLC-01287 Pre-Cleaned Square Wide-Mouth Bottle, Clear Glass, 60mL; 48/CS EW-34610-20 Mfr # GLC-01287
COMPARE
60 28 mm Glass
$178.00USD / Case of 48
Cole-Parmer Essentials GLC-01314 Pre-Cleaned Square Wide-Mouth Bottle, Clear Glass, 125mL; 24/CS EW-34610-30 Mfr # GLC-01314
COMPARE
125 33 mm Glass
$125.00USD / Case of 24
Cole-Parmer Essentials GLC-01339 Pre-Cleaned Square Wide-Mouth Bottle, Clear Glass, 250mL; 24/CS EW-34610-40 Mfr # GLC-01339
COMPARE
250 43 mm Glass
$169.00USD / Case of 24
Cole-Parmer Essentials GLC-01367 Pre-Cleaned Square Wide-Mouth Bottle, Clear Glass, 500mL; 24/CS EW-34610-50 Mfr # GLC-01367
COMPARE
500 48 mm Glass
$197.00USD / Case of 24
Cole-Parmer Essentials GLC-01391 Pre-Cleaned Square Wide-Mouth Bottle, Clear Glass, 1000mL; 12/CS EW-34610-60 Mfr # GLC-01391
COMPARE
1000 58 mm Glass
$160.00USD / Case of 12
Showing 1- 7 of 7