Quality Process Gauge
Flutterless movement gives filled-like readings at lower cost
  • Screw-on lens makes the gauge easy to service and calibrate without removing gauge from process line
Process gauges are versatile instruments that come standard with a screw-on acrylic lens and recalibrator screw. Pressure gauge is also equipped with a flutterless movement for added stability. Gauges have phosphor bronze Bourdon tube, brass socket, black steel case, and polycarbonate lens.
$20.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Winters Instruments Q90211 Quality Process Gauge, 0-15 psi - 2 1/2" dial EW-68612-02 Mfr # P9S90211
COMPARE
±2-1-2% (ANSI/ASME Grade B) 15
$20.00USD / Each