Quick-Disconnect Couplings for Air Lines

  • Coupling options are available in both 1⁄4" and 3 ⁄8" standard connections
$2.20 - $5.40USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Port Size
Availability Pricing
Puma MS-1/4F Quick Disconnect Air Coupler, 1/4 in NPT(F) EW-07123-38 Mfr # MS-1/4F
COMPARE
1/4" NPT(F)
$5.10USD / Each
Puma MS-3/8F Quick Disconnect Air Coupler, 3/8 in NPT(F) EW-07123-39 Mfr # MS-3/8F
COMPARE
3/8" NPT(F)
$5.40USD / Each
Puma MS-1/4M Quick Disconnect Air Coupler, 1/4 in NPT(M) EW-07123-40 Mfr # MS-1/4M
COMPARE
1/4" NPT(F)
$5.10USD / Each
Puma MP-1/4F Quick Disconnect Air Plug, 1/4 in NPT(F) EW-07123-42 Mfr # MP-1/4F
COMPARE
1/4" NPT(F)
$2.20USD / Each
Puma MP-3/8F Quick Disconnect Air Plug, 3/8 in NPT(F) EW-07123-43 Mfr # MP-3/8F
COMPARE
3/8" NPT(F)
$2.75USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat