Quick-Disconnect Male Pipe Thread Inserts
High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately). Plug insert into coupling body to connect—fitting halves lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 SS latch springs and EPDM seals.
$7.90 - $103.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Quick-Disconnect Flow Size (in) Thread size NPT(M) (port 1) (in)
Availability Pricing
CPC (Colder) HFC24612 Quick-disconnect fittings, Straight-through Male pipe thread inserts, PP, 3/8"
Buy More Save More
EW-31303-30 Mfr # HFC24612
COMPARE
Polypropylene 3/8 3/8
$7.90USD / Each
CPC (Colder) HFC24812 Quick-disconnect fittings, Straight-through Male pipe thread inserts, PP, 1/2"
Buy More Save More
EW-31303-31 Mfr # HFC24812
COMPARE
Polypropylene 3/8 1/2
$7.90USD / Each
CPC (Colder) HFC241212 Quick-disconnect fittings, Straight-through Male pipe thread inserts, PP, 3/4"
Buy More Save More
EW-31303-32 Mfr # HFC241212
COMPARE
Polypropylene 3/8 3/4
$8.40USD / Each
CPC (Colder) HFCD24612 Quick-disconnect fittings, Valved Male pipe thread inserts, PP, 3/8"
Buy More Save More
EW-31303-35 Mfr # HFCD24612
COMPARE
Polypropylene 3/8 3/8
$14.50USD / Each
CPC (Colder) HFCD24812 Quick-disconnect fittings, Valved Male pipe thread inserts, PP, 1/2"
Buy More Save More
EW-31303-36 Mfr # HFCD24812
COMPARE
Polypropylene 3/8 1/2
$14.30USD / Each
CPC (Colder) HFCD241212 Quick-disconnect fittings, Valved Male pipe thread inserts, PP, 3/4"
Buy More Save More
EW-31303-37 Mfr # HFCD241212
COMPARE
Polypropylene 3/8 3/4
$15.80USD / Each
CPC (Colder) HFC24835 Quick-disconnect fittings, straight-through male pipe thread inserts, PES, 1/2"
Buy More Save More
EW-31307-31 Mfr # HFC24835
COMPARE
Polysulfone 3/8 1/2
$10.40USD / Each
CPC (Colder) HFC241235 Quick-disconnect fittings, straight-through male pipe thread inserts, PES, 3/4"
Buy More Save More
EW-31307-32 Mfr # HFC241235
COMPARE
Polysulfone 3/8 3/4
$11.10USD / Each
CPC (Colder) HFCD24635 Quick-disconnect fittings, valved male pipe thread inserts, PES, 3/8"
Buy More Save More
EW-31307-35 Mfr # HFCD24635
COMPARE
Polysulfone 3/8 3/8
$18.50USD / Each
CPC (Colder) HFCD24835 Quick-disconnect fittings, valved male pipe thread inserts, PES, 1/2"
Buy More Save More
EW-31307-36 Mfr # HFCD24835
COMPARE
Polysulfone 3/8 1/2
$18.50USD / Each
CPC (Colder) HFCD241235 Quick-disconnect fittings, valved male pipe thread inserts, PES, 3/4"
Buy More Save More
EW-31307-37 Mfr # HFCD241235
COMPARE
Polysulfone 3/8 3/4
$18.70USD / Each
CPC (Colder) HFC24635 Quick-disconnect fittings, straight-through male pipe thread inserts, PES, 3/8"; 10/PK EW-31308-72 Mfr # HFC24635
COMPARE
Polysulfone 3/8 3/8
$103.00USD / Pkg of 10
Showing 1- 12 of 12