CPC® Quick-Disconnect Fittings, Sanitary Bodies and Inserts
Attach directly to your sanitary hardware assemblies!
  • USP Class IV materials
  • Fully sterilizable
These couplings allow you to integrate disposables with your existing stainless steel processing equipment. Both couplings and inserts are available with standard sanitary terminations to attach to your system with a sanitary gasket and tri-clover clamps. All models feature USP Class VI platinum-cured silicone gaskets.
These quick-disconnect couplings require a large fitting body and large insert (order separately).
$16.40 - $237.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in) Description
Availability Pricing
CPC (Colder) Quick-Disconnect Fitting, Sanitary Connector Body, Polysulfone, 3/4" Sanitary; 1/Ea EW-31604-26 Mfr # MPX3301239
COMPARE
Polysulfone 0.75 Quick-Disconnect Fitting, Sanitary Connector Body, Polysulfone, 3/4" Sanitary; 1/Ea
$17.00USD / Each
CPC (Colder) Quick-Disconnect Fitting, Sanitary Connector Body, Polysulfone, 3/4" Sanitary; 1/Ea EW-31604-27 Mfr # MPC3301239
COMPARE
Polysulfone 0.75 Quick-Disconnect Fitting, Sanitary Connector Body, Polysulfone, 3/4" Sanitary; 1/Ea
$16.40USD / Each
CPC (Colder) Quick-Disconnect Fitting, Sanitary Connector Body, Polysulfone, 1" Sanitary; 1/Ea EW-31604-28 Mfr # MPC3301639
COMPARE
Polysulfone 1 Quick-Disconnect Fitting, Sanitary Connector Body, Polysulfone, 1" Sanitary; 1/Ea
$18.10USD / Each
CPC (Colder) Quick-Disconnect Fitting, Sanitary Connector Inert, Stainless Steel, 3/4" Sanitary; 1/Ea EW-31604-30 Mfr # SQCC221212M
COMPARE
Stainless Steel 0.75 Quick-Disconnect Fitting, Sanitary Connector Inert, Stainless Steel, 3/4" Sanitary; 1/Ea
$237.00USD / Each
CPC (Colder) Quick-Disconnect Fitting, Sanitary Connector Inert, Stainless Steel, 1" Sanitary; 1/Ea EW-31604-32 Mfr # SQCX222416M
COMPARE
Stainless Steel 1 Quick-Disconnect Fitting, Sanitary Connector Inert, Stainless Steel, 1" Sanitary; 1/Ea
$237.00USD / Each
CPC (Colder) Quick-Disconnect Fitting, Sanitary Connector Inert, Stainless Steel, 1-1/2" Sanitary; 1/Ea EW-31604-34 Mfr # SQCX222424M
COMPARE
Stainless Steel 1.5 Quick-Disconnect Fitting, Sanitary Connector Inert, Stainless Steel, 1-1/2" Sanitary; 1/Ea
$237.00USD / Each