ThermoTek Rack Mount Thermoelectric Chillers
Solid state reliability for superior performance and temperature stability
  • Quiet operation, low vibration
  • 3U tall with integrated front handles
  • Leak-free disconnects
  • Touch-screen interface with USB and RS-232 connectivity
Introducing the industry’s smallest rack mountable recirculating chiller at just 3U tall. These ThermoTek thermoelectric chillers are built with solid state reliability for superior performance and temperature stability. The chillers are designed to maintain a set temperature of the working fluid that is circulated between the thermal management application and itself. Standard features include fan speed control for reduced noise, touch-screen interface, and connectivity with RS-232 and USB.
Choose from models with a centrifugal or gear pump and from models with standard only or standard and advanced features including flow meter, remote temperature sensing capabilities, and electrical interlock.
$5,034.00 - $8,514.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Cooling Capacity At 0° C (watts) Depth (in) Max Pump Pressure (PSI)
Availability Pricing
ThermoTek 3U Rack Mount Thermoelectric Chiller, 22", Centrifugal Pump, 240 Watts; 120-240 VAC EW-12751-97 Mfr # 0P9RC22A25020R0-XSC
COMPARE
240 22 15
$5,034.00USD / Each
ThermoTek 3U Rack Mount Thermoelectric Chiller, 22", Centrifugal Pump, 485 Watts; 120-240 VAC EW-12751-98 Mfr # 0P9RC22A50020R0-XSC
COMPARE
485 22 15
$6,477.00USD / Each
ThermoTek 3U Rack Mount Thermoelectric Chiller, 22", Centrifugal Pump, 700 Watts; 120-240 VAC EW-12751-99 Mfr # 0P9RC22A75020R0-XSC
COMPARE
700 22 15
$7,457.00USD / Each
ThermoTek 3U Rack Mount Thermoelectric Chiller, with Flow Meter and Remote Control, 15", Centrifugal Pump, 240 Watts; 120-240 VAC EW-12752-00 Mfr # 0P9RC15A25020R0-FSC
COMPARE
240 15 15
$5,575.00USD / Each
ThermoTek 3U Rack Mount Thermoelectric Chiller, with Flow Meter and Remote Control, 22", Centrifugal Pump, 240 Watts; 120-240 VAC EW-12752-01 Mfr # 0P9RC22A25020R0-FSC
COMPARE
240 22 15
$5,324.00USD / Each
ThermoTek 3U Rack Mount Thermoelectric Chiller, with Flow Meter and Remote Control, 22", Gear Pump, 240 Watts; 120-240 VAC EW-12752-02 Mfr # 0P9RC22A25050R0-FSC
COMPARE
240 22 45.3
$6,236.00USD / Each
ThermoTek 3U Rack Mount Thermoelectric Chiller, with Flow Meter and Remote Control, 22", Gear Pump, 485 Watts; 120-240 VAC EW-12752-03 Mfr # 0P9RC22A50050R0-FSC
COMPARE
485 22 45.3
$7,656.00USD / Each
ThermoTek 3U Rack Mount Thermoelectric Chiller, with Flow Meter and Remote Control, 22", Centrifugal Pump, 700 Watts; 120-240 VAC EW-12752-04 Mfr # 0P9RC22A75020R0-FSC
COMPARE
700 22 15
$7,673.00USD / Each
ThermoTek 3U Rack Mount Thermoelectric Chiller, with Flow Meter and Remote Control, 22", Gear Pump, 700 Watts; 120-240 VAC EW-12752-05 Mfr # 0P9RC22A75050R0-FSC
COMPARE
700 22 45.3
$8,514.00USD / Each
Showing 1- 9 of 9