RADNOR® SmoothDome™ Polyethylene Hard Hat with Ratchet Suspension
Sturdy, lightweight polyethylene shell provides safety and comfort all day
  • Dielectric properties offer protection from electrical charge
  • Ratchet suspension absorbs shock and redirects impact force away from head
  • Meets ANSI Type 1, Class E standards
These sturdy, lightweight polyethylene slotted cap-style hard hats provide a comfortable fit all day long. The smooth silhouette design can easily accommodate welders’ shields. The adjustable ratchet suspension allows for a custom fit.
$22.70USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Material Description
Availability Pricing
Radnor® White SmoothDome™ Polyethylene Cap-Style Hard Hat with Ratchet Suspension EW-78900-51 Mfr # RAD64051020
COMPARE
White Polyethylene(PE) White SmoothDome™ Polyethylene Cap Style Hard Hat with Ratchet Suspension
$22.70USD / Each
Radnor® Hi-Viz Yellow SmoothDome™ Polyethylene Cap-Style Hard Hat with Ratchet Suspension EW-78900-52 Mfr # RAD64051026
COMPARE
Yellow Polyethylene(PE) Hi-Viz Yellow SmoothDome™ Polyethylene Cap Style Hard Hat with Ratchet Suspension
$22.70USD / Each