This product is now discontinued, View Related Products

Rainwise Dual Display Rain Gauges

Meets National Weather Service (NWS) specifications for statistical accuracy
  • Monitor current and cumulative rainfall
These self-emptying, tipping bucket rain gauges monitor both current and cumulative rainfall levels. Each tip of the bucket transmits a 0.01” register on the LCDs. Reset the display measurements to zero after each rainfall with the push of a button. Select from cabled or wireless rain gauges to fit your application.

Cabled rain gauge (99752-04) meet the National Weather Service (NWS) specifications for statistical accuracy. System requires two AA batteries (not included) for display power.

Wireless rain gauge (99753-12) transmits rainfall measurements up to 300 feet away. Rain collector requires two AA batteries (not included) and the LCD requires three D batteries (not included). Order batteries separately.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description Rain Fall Accuracy (in)
Availability Pricing
Rainwise RAINEW-211 Cabled Dual Display Rain Gauge EW-99752-04 Mfr # RAINEW-211
COMPARE
Cabled Dual Display Rain Gauge ±2% at 1.5\" per hour
Call For Price
Rainwise RAINEW-212 Dual Display Rain Gauge, 30ft cable EW-99752-06 Mfr # RAINEW-212
COMPARE
Dual Display Rain Gauge, 30ft cable
Call For Price
Rainwise RAINEW-WIRELESS Wireless Dual Display Rain Gauge EW-99753-12 Mfr # RAINEW-WIRELESS
COMPARE
Wireless Dual Display Rain Gauge ±2% at 1.5\" per hour
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat