Ralston Pressure Only and Pressure/Vacuum Hand Pumps
Create pressure or vaccum and calibrate instrumentation using a pressure gauge or calibrator attached to the pump
Pressure-only hand pumps
  • Light weight, compact pump generates up to 125 psi (8.6 bar) with ease
  • Large volume fine adjustment piston provides precise control, even at very low pressures
  • Modular design makes field maintenance quick and easy
  • Precisely vent pressure with soft seated bleed valve
  • Two outlet ports provide connections for both pressure reference and for device under test thus eliminating a tee
  • Low volume Quick-test hose, gauge adapter and process connection can be attached to pump without thread sealant or a wrench
  • Rugged, all metal construction reduces pressure changes due to temperature fluctuations
  • Pressure and vacuum hand pumps
  • Unique design allows the user to convert from pressure to vacuum by simply twisting a knob, even with gloves on
  • Two outlet ports allow connection to reference pressure gauge or pressure calibrator as well as device under test, thus eliminating a tee
  • Large volume fine adjustment piston provides precise control, even at very low pressures
    Precisely vent pressure with soft seated bleed valve
  • $502.00 - $1,230.00USD / Each
    10 variations of this product are available.
    See All Product Options Below
    10 PRODUCT OPTIONS
    Compareitem specifications
    Max Pressure Pressure Resolution Description
    Availability Pricing
    Ralston Instruments DP0V-0000 Pneumatic Hand Pump, 125 psi EW-68976-60 Mfr # DP0V-0000
    COMPARE
    125 psi (7 bar) ±0.01 psi Pneumatic Hand Pump, 125 psi
    $502.00USD / Each
    Ralston Instruments DP0V-1MBA Pneumatic Hand Pump, 125 psi, 1/8"NPT(M) adapter EW-68976-61 Mfr # DP0V-1MBA
    COMPARE
    125 psi (7 bar) ±0.01 psi Pneumatic Hand Pump, 125 psi, 1/8"NPT(M) adapter
    $525.00USD / Each
    Ralston Instruments DP0V-2MBA Pneumatic Hand Pump, 125 psi, 1/4"NPT(M) adapter EW-68976-62 Mfr # DP0V-2MBA
    COMPARE
    125 psi (7 bar) ±0.01 psi Pneumatic Hand Pump, 125 psi, 1/4"NPT(M) adapter
    $522.00USD / Each
    Ralston Instruments DP0V-30PSIG-D Pneumatic Hand Pump, 125 psi, 1/4"NPT(F), Digital EW-68976-65 Mfr # DP0V-30PSIG-D
    COMPARE
    125 psi (7 bar) ±0.01 psi Pneumatic Hand Pump, 125 psi, 1/4"NPT(F), Digital
    $1,080.00USD / Each
    Ralston Instruments DP0V-100PSIG-D Pressure Handpump DP0V-100psig-d EW-68976-67 Mfr # DP0V-100PSIG-D
    COMPARE
    125 psi (7 bar) ±0.01 psi Pressure Handpump DP0V-100psig-d
    $1,100.00USD / Each
    Ralston Instruments DPPV-0000 Pressure/vacuum Handpump EW-68976-68 Mfr # DPPV-0000
    COMPARE
    25 inHg (-635 mmHg) ±0.01 psi Pressure/vacuum Handpump
    $710.00USD / Each
    Ralston Instruments DPPV-1MBA Pressure/vacuum Handpump Dppv-1mba EW-68976-69 Mfr # DPPV-1MBA
    COMPARE
    25 inHg (-635 mmHg) ±0.01 psi Pressure/vacuum Handpump Dppv-1mba
    $696.00USD / Each
    Ralston Instruments DPPV-2MBA Pressure/vacuum Handpump Dppv-2mba EW-68976-70 Mfr # DPPV-2MBA
    COMPARE
    25 inHg (-635 mmHg) ±0.01 psi Pressure/vacuum Handpump Dppv-2mba
    $721.00USD / Each
    Ralston Instruments DPPV-2FBA Pressure/vacuum Handpump Dppv-2fba EW-68976-71 Mfr # DPPV-2FBA
    COMPARE
    25 inHg (-635 mmHg) ±0.01 psi Pressure/vacuum Handpump Dppv-2fba
    $715.00USD / Each
    Ralston Instruments DPPV-15PSIG-D Pressure/vacuum Handpump Dppv-15psig-d EW-68976-72 Mfr # DPPV-PM15PSIG-D
    COMPARE
    25 inHg (-635 mmHg) ±0.01 psi Pressure/vacuum Handpump Dppv-15psig-d
    $1,230.00USD / Each
    Showing 1- 10 of 10
    Need Help with this product?
    Call Us At1-847-549-7600
    Live chat