Reagents® Chloroform, Purified

Manufactured using the highest quality raw materials in ISO 9001 facility
$52.00 - $733.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Chloroform, 500mL, Purified EW-78915-09 Mfr # C2175500-500C
COMPARE
Solvents 500 Chloroform, 500mL, Purified
$52.00USD / Each
Reagents Chloroform, 1L, Purified EW-78918-60 Mfr # C2175500-1C
COMPARE
Solvents 1000 Chloroform, 1L, Purified
$86.00USD / Each
Reagents Chloroform, 4L, Purified EW-78925-49 Mfr # C2175500-4C
COMPARE
Solvents 4000 Chloroform, 4L, Purified
$211.00USD / Each
Reagents Chloroform, 20L, Purified EW-78929-85 Mfr # C2175500-20E6
COMPARE
Solvents 20000 Chloroform, 20L, Purified
$733.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat