Reagents® Deionized Water, ACS Reagent, ASTM Type I

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Water meets ASTM Type I D 1193 reagent grade certification
  • Colorless liquid
  • Density: 1
  • Solubility information: miscible
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$13.33 - $2,214.00USD / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Packaging Purity Grade Description
Availability Pricing
Reagents Deionized Water, ACS Reagent, ASTM Type I (D 1193), Cubitainer® Package, 10 L
New Products
EW-62286-25 Mfr # C2346500-10F
COMPARE
Cubitainer® Package ACS Reagent Deionized Water, ACS Reagent, ASTM Type I (D 1193), Cubitainer® Package, 10 L
$48.00USD / Each
Reagents Deionized Water, ACS Reagent, ASTM Type I (D 1193), Poly Bottle, 1 L
New Products
EW-62286-26 Mfr # C2346500-1A
COMPARE
Poly Bottle ACS Reagent Deionized Water, ACS Reagent, ASTM Type I (D 1193), Poly Bottle, 1 L
$21.17USD / Each
Reagents Deionized Water, ACS Reagent, ASTM Type I (D 1193), Tote, 275 gal.
New Products
EW-62286-27 Mfr # C2346500-275E3
COMPARE
Tote ACS Reagent Deionized Water, ACS Reagent, ASTM Type I (D 1193), Tote, 275 gal.
$2,214.00USD / Each
Reagents Deionized Water, ACS Reagent, ASTM Type I (D 1193), Poly Bottle, 500 mL
New Products
EW-62286-28 Mfr # C2346500-500A
COMPARE
Poly Bottle ACS Reagent Deionized Water, ACS Reagent, ASTM Type I (D 1193), Poly Bottle, 500 mL
$13.33USD / Each
Reagents Deionized Water, ACS Reagent, ASTM Type I (D 1193), Drum, 55 gal.
New Products
EW-62286-29 Mfr # C2346500-55E
COMPARE
Drum ACS Reagent Deionized Water, ACS Reagent, ASTM Type I (D 1193), Drum, 55 gal.
$485.80USD / Each
Reagents Deionized Water, ACS Reagent, ASTM Type I (D 1193), Natural Poly Bottle, 4 L EW-78922-50 Mfr # C2346500-4A
COMPARE
Poly bottle ACS Reagent Deionized Water, ACS Reagent, ASTM Type I (D 1193), Natural Poly Bottle, 4 L
$40.80USD / Each
Reagents Deionized Water, ACS Reagent, ASTM Type I (D 1193), Cubitainer® Package, 20 L EW-78927-72 Mfr # C2346500-20F
COMPARE
Cubitainer ACS Reagent Deionized Water, ACS Reagent, ASTM Type I (D 1193), Cubitainer® Package, 20 L
$95.80USD / Each
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat