Reagents® Hydrochloric Acid, 1.0N, Standardized
Manufactured using the highest quality raw materials in ISO 9001 facility
  • N/1, Assay 0.998 to 1.002 Normal, Aqueous
Hydrochloric Acid, 1L, 1N ConcentratIon, Colorless, Liquid, 100°C Boiling Point, 0°C Melting Point, <1 pH Range, 1.02 Density, 24 Months Shelf Life.
$27.50 - $187.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Hydrochloric Acid, 1.0N, 500mL, Standardized EW-78911-56 Mfr # C7101080-500A
COMPARE
Standardized 500 Hydrochloric Acid, 1.0N, 500mL, Standardized
$27.50USD / Each
Reagents Hydrochloric Acid, 1.0N, 1L, Standardized EW-78915-73 Mfr # C7101080-1A
COMPARE
Standardized 1000 Hydrochloric Acid, 1.0N, 1L, Standardized
$31.30USD / Each
Reagents Hydrochloric Acid, 1.0N, 4L, Standardized, Natural Poly Container EW-78921-46 Mfr # C7101080-4A
COMPARE
Standardized 4000 Hydrochloric Acid, 1.0N, 4L, Standardized
$66.40USD / Each
Reagents Hydrochloric Acid, 1.0N, 4L, Standardized, Cubitainer EW-78921-47 Mfr # C7101080-4F
COMPARE
Standardized 4000 Hydrochloric Acid, 1.0N, 4L, Standardized
$65.30USD / Each
Reagents Hydrochloric Acid, 1.0N, 10L, Standardized EW-78925-84 Mfr # C7101080-10F
COMPARE
Standardized 10000 Hydrochloric Acid, 1.0N, 10L, Standardized
$145.25USD / Each
Reagents Hydrochloric Acid, 1.0N, 20L, Standardized, Cubitainer EW-78928-07 Mfr # C7101080-20F
COMPARE
Standardized 20000 Hydrochloric Acid, 1.0N, 20L, Standardized
$187.00USD / Each