Reagents® Hydrochloric Acid, 37%, ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 36.5 to 38%
  • Density: 1.18
  • Boiling point: approximately 100 °C (212 °F)
  • Solubility information: miscible
  • pH range: <1
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$24.00 - $316.25USD / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
Reagents Hydrochloric Acid, 37%, ACS Reagent, Amber Glass Bottle, 1 L
New Products
EW-62286-36 Mfr # C5101100-1C
COMPARE
Acid Amber glass bottle 1000
$44.31USD / Each
Reagents Hydrochloric Acid, 37%, ACS Reagent, Poly Bottle, 2.5 L
New Products
EW-62286-37 Mfr # C5101100-2.5A
COMPARE
Acid Poly bottle 2500
$70.00USD / Each
Reagents Hydrochloric Acid, 37%, ACS Reagent, Poly Bottle, 500 mL
New Products
EW-62286-38 Mfr # C5101100-500A
COMPARE
Acid Poly bottle 500
$24.00USD / Each
Reagents Hydrochloric Acid, 37%, ACS Reagent, Amber Glass Bottle, 500mL
New Products
EW-62286-39 Mfr # C5101100-500C
COMPARE
Acid Amber glass bottle 500
$29.00USD / Each
Reagents Hydrochloric Acid, 37%, ACS Reagent, Winpak Container, 20 L EW-78929-53 Mfr # C5101100-20E7
COMPARE
Acid Winpak container 20000
$316.25USD / Each
Reagents Hydrochloric Acid, 37%, ACS Reagent, Clear Glass Bottle, 2.5 L EW-78930-10 Mfr # C5101100-2.5D1
COMPARE
Acid Clear glass bottle 2500
$98.10USD / Each
Reagents Hydrochloric Acid, 37%, ACS Reagent, Natural Poly Bottle, 4 L EW-78930-11 Mfr # C5101100-4A
COMPARE
Acid Poly bottle 4000
$156.75USD / Each
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat