Reagents® Hydrochloric Acid 50% (v/v), Volumetric
Manufactured using the highest quality raw materials in ISO 9001 facility
  • 1+1
Hydrochloric Acid, 4L, 50% v/v ConcentratIon, Colorless to Slightly Greenish-yellow, Liquid, 100°C Boiling Point, 0°C Melting Point, <1 pH Range, 1.1 Density, 37 Months Shelf Life.
$49.00 - $472.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Hydrochloric Acid, 50% (v/v), 500mL, Volumetric EW-78911-58 Mfr # C5505000-500A
COMPARE
Volumetric 500 Hydrochloric Acid, 50% (v/v), 500mL, Volumetric
$49.00USD / Each
Reagents Hydrochloric Acid, 50% (v/v), 1L, Volumetric EW-78915-87 Mfr # C5505000-1A
COMPARE
Volumetric 1000 Hydrochloric Acid, 50% (v/v), 1L, Volumetric
$66.00USD / Each
Reagents Hydrochloric Acid, 50% (v/v), 2.5L, Volumetric EW-78919-66 Mfr # C5505000-2.5D1
COMPARE
Volumetric 2500 Hydrochloric Acid, 50% (v/v), 2.5L, Volumetric
$93.00USD / Each
Reagents Hydrochloric Acid, 50% (v/v), 4L, Volumetric EW-78923-02 Mfr # C5505000-4A
COMPARE
Volumetric 4000 Hydrochloric Acid, 50% (v/v), 4L, Volumetric
$166.00USD / Each
Reagents Hydrochloric Acid, 50% (v/v), 10L, Volumetric EW-78926-45 Mfr # C5505000-10F
COMPARE
Volumetric 10000 Hydrochloric Acid, 50% (v/v), 10L, Volumetric
$236.00USD / Each
Reagents Hydrochloric Acid, 50% (v/v), 20L, Volumetric EW-78929-22 Mfr # C5505000-20E7
COMPARE
Volumetric 20000 Hydrochloric Acid, 50% (v/v), 20L, Volumetric
$472.00USD / Each