Reagents® Hydrochloric Acid, 6.0N, Standardized

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Assay 5.95 to 6.05 Normal, Aqueous
Hydrochloric Acid, 1L, 6N ConcentratIon, Colorless, Liquid, 100°C Boiling Point, 0°C Melting Point, <1 pH Range, 1.1 Density, 24 Months Shelf Life.
$52.00 - $452.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Hydrochloric Acid, 6.0N, 1L, Standardized EW-78915-88 Mfr # C7101150-1A
COMPARE
Standardized 1000
$52.00USD / Each
Reagents Hydrochloric Acid, 6.0N, 4L, Standardized EW-78923-03 Mfr # C7101150-4A
COMPARE
Standardized 4000
$152.00USD / Each
Reagents Hydrochloric Acid, 6.0N, 10L, Standardized EW-78926-46 Mfr # C7101150-10F
COMPARE
Standardized 10000
$279.00USD / Each
Reagents Hydrochloric Acid, 6.0N, 20L, Standardized EW-78929-23 Mfr # C7101150-20E7
COMPARE
Standardized 20000
$452.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat