This product is now discontinued, View Related Products
Reagents® Nitric Acid 70% Trace Metal, ACS
Manufactured using the highest quality raw materials in ISO 9001 facility
  • 68.00-70.00%
Nitric Acid, 2.5L, 70% Concentration, Trace Metal Grade, Colorless, Liquid, 121°C Boiling Point, -32°C Melting Point, <1 pH Range, 1.416 Density, 37 Months Shelf Life.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Nitric Acid 70% Trace Metal, 2.5L , ACS EW-78930-13 Mfr # C5101650-2.5D
COMPARE
Acid 2500 Nitric Acid 70% Trace Metal, 2.5L, ACS
Call For Price
Reagents Nitric Acid 70% Trace Metal, SC, 2.5L , ACS EW-78930-14 Mfr # C5101650-2.5D1
COMPARE
Acid 2500 Nitric Acid 70% Trace Metal, SC, 2.5L, ACS